Anti-TUBB1 Mouse Monoclonal Antibody [clone: OTI1G5]

Lieferant: OriGene
TA506622
ORIGTA506622EA 621 EUR
ORIGTA506622
Anti-TUBB1 Mouse Monoclonal Antibody [clone: OTI1G5]
Antikörper
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: TUBB1 (tubulin, beta 1 class VI)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR