Anti-EPCAM Mouse Monoclonal Antibody [clone: OTI1C1]

Lieferant: OriGene
TA506629
ORIGTA506629EA 621 EUR
ORIGTA506629
Anti-EPCAM Mouse Monoclonal Antibody [clone: OTI1C1]
Antikörper
Recommended Dilutions: Western Blot: 1:400-1:1000; Immunochemistry: 1:150-1:500; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: EPCAM (epithelial cell adhesion molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR