Anti-EPCAM Mouse Monoclonal Antibody [clone: OTI2B6]

Lieferant: OriGene
TA506643
ORIGTA506643EA 621 EUR
ORIGTA506643
Anti-EPCAM Mouse Monoclonal Antibody [clone: OTI2B6]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: EPCAM (epithelial cell adhesion molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR