Anti-FOXP1 Mouse Monoclonal Antibody [clone: OTI4D2]

Lieferant: OriGene
TA506700
ORIGTA506700EA 621 EUR
ORIGTA506700
Anti-FOXP1 Mouse Monoclonal Antibody [clone: OTI4D2]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50-1:150; Immunofluorescence: 1:50-1:100

Type: Primary
Antigen: FOXP1 (forkhead box P1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR