Anti-CD44 Mouse Monoclonal Antibody [clone: OTI1A2]

Lieferant: OriGene
TA506729
ORIGTA506729EA 621 EUR
ORIGTA506729
Anti-CD44 Mouse Monoclonal Antibody [clone: OTI1A2]
Antikörper
Recommended Dilutions: Western Blot: 1:8000; Immunofluorescence: 1:100; Flow Cytometry: 1:50

Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR