Anti-CD44 Mouse Monoclonal Antibody [clone: 2E3]

Lieferant: OriGene
TA506834
ORIGTA506834EA 621 EUR
ORIGTA506834
Anti-CD44 Mouse Monoclonal Antibody [clone: 2E3]
Antikörper
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 2E3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR