Anti-SIVA1 Mouse Monoclonal Antibody [clone: OTI2D3]

Lieferant: OriGene
TA506835
ORIGTA506835EA 621 EUR
ORIGTA506835
Anti-SIVA1 Mouse Monoclonal Antibody [clone: OTI2D3]
Antikörper
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: SIVA1 (SIVA1, apoptosis-inducing factor)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR