Anti-PAK7 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA590312
ORIGTA590312EA 669 EUR
ORIGTA590312
Anti-PAK7 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:5000-1:20000; ELISA: 1:100-1:2000

Type: Primary
Antigen: PAK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR