Anti-SPR Mouse Monoclonal Antibody [clone: OTI3D6]

Lieferant: OriGene
TA600129
ORIGTA600129EA 636 EUR
ORIGTA600129
Anti-SPR Mouse Monoclonal Antibody [clone: OTI3D6]
Antikörper
Recommended Dilutions: 1:100-1:1000

Type: Primary
Antigen: SPR (sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR