Anti-TBXAS Mouse Monoclonal Antibody [clone: OTI2C1]

Lieferant: OriGene
TA600231
ORIGTA600231EA 636 EUR
ORIGTA600231
Anti-TBXAS Mouse Monoclonal Antibody [clone: OTI2C1]
Antikörper
Recommended Dilutions: 1:100-1:1000

Type: Primary
Antigen: TBXAS
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR