Anti-TIMP2 Mouse Monoclonal Antibody [clone: 1D7]

Lieferant: OriGene
TA600237
ORIGTA600237EA 636 EUR
ORIGTA600237
Anti-TIMP2 Mouse Monoclonal Antibody [clone: 1D7]
Antikörper
Recommended Dilutions: 1:100-1:1000

Type: Primary
Antigen: TIMP2 (TIMP metallopeptidase inhibitor 2)
Clonality: Monoclonal
Clone: 1D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR