Anti-PECAM1 Mouse Monoclonal Antibody [clone: OTI4D1]

Lieferant: OriGene
TA600248
ORIGTA600248EA 636 EUR
ORIGTA600248
Anti-PECAM1 Mouse Monoclonal Antibody [clone: OTI4D1]
Antikörper
Recommended Dilutions: 1:100-1:1000

Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR