Anti-MEF2C Mouse Monoclonal Antibody [clone: OTI1B6]

Lieferant: OriGene
TA700065
ORIGTA700065EA 686 EUR
ORIGTA700065
Anti-MEF2C Mouse Monoclonal Antibody [clone: OTI1B6]
Antikörper
Recommended Dilutions: 1:250-1:1000

Type: Primary
Antigen: MEF2C (myocyte enhancer factor 2C)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR