Anti-DTNB Mouse Monoclonal Antibody [clone: OTI2E3]

Lieferant: OriGene
TA700289
ORIGTA700289EA 686 EUR
ORIGTA700289
Anti-DTNB Mouse Monoclonal Antibody [clone: OTI2E3]
Antikörper
  • Recommended Dilutions: 1:250-1:1000

Type: Primary
Antigen: DTNB (dystrobrevin, beta)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR