Anti-SERPINB3 Mouse monoclonal Antibody (Biotin) [clone: 2H2]

Lieferant: OriGene
TA700493
ORIGTA700493EA 686 EUR
ORIGTA700493
Anti-SERPINB3 Mouse monoclonal Antibody (Biotin) [clone: 2H2]
Antikörper
Recommended Dilutions: 1:250-1:1000

Type: Primary
Antigen: SERPINB3
Clonality: monoclonal
Clone: 2H2
Conjugation: Biotin
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR