Anti-MIEN1 Mouse Monoclonal Antibody [clone: OTI3H9]

Lieferant: OriGene
TA800018
ORIGTA800018EA 621 EUR
ORIGTA800018
Anti-MIEN1 Mouse Monoclonal Antibody [clone: OTI3H9]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: MIEN1 (migration and invasion enhancer 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR