Anti-DPPA4 Mouse Monoclonal Antibody [clone: OTI1E1]

Lieferant: OriGene
TA800032
ORIGTA800032EA 621 EUR
ORIGTA800032
Anti-DPPA4 Mouse Monoclonal Antibody [clone: OTI1E1]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: DPPA4 (developmental pluripotency associated 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR