Anti-ZNF365 Mouse Monoclonal Antibody [clone: OTI3B1]

Lieferant: OriGene
TA800111
ORIGTA800111EA 621 EUR
ORIGTA800111
Anti-ZNF365 Mouse Monoclonal Antibody [clone: OTI3B1]
Antikörper
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ZNF365 (zinc finger protein 365)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR