Anti-MLF1 Mouse Monoclonal Antibody [clone: OTI2C10]

Lieferant: OriGene
TA800121
ORIGTA800121EA 621 EUR
ORIGTA800121
Anti-MLF1 Mouse Monoclonal Antibody [clone: OTI2C10]
Antikörper
  • Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: MLF1 (myeloid leukemia factor 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR