Anti-FOXI1 Mouse Monoclonal Antibody [clone: OTI2G6]

Lieferant: OriGene
TA800146
ORIGTA800146EA 621 EUR
ORIGTA800146
Anti-FOXI1 Mouse Monoclonal Antibody [clone: OTI2G6]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: FOXI1 (forkhead box I1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR