Anti-SF3A1 Mouse Monoclonal Antibody [clone: OTI1B5]

Lieferant: OriGene
TA800496
ORIGTA800496EA 621 EUR
ORIGTA800496
Anti-SF3A1 Mouse Monoclonal Antibody [clone: OTI1B5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SF3A1 (splicing factor 3a, subunit 1, 120kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR