Anti-CBX8 Mouse Monoclonal Antibody [clone: OTI2C8]

Lieferant: OriGene
TA800500
ORIGTA800500EA 621 EUR
ORIGTA800500
Anti-CBX8 Mouse Monoclonal Antibody [clone: OTI2C8]
Antikörper
  • Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CBX8 (chromobox homolog 8)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR