Anti-CD4 Mouse Monoclonal Antibody [clone: OTI5D9]

Lieferant: OriGene
TA800512
ORIGTA800512EA 621 EUR
ORIGTA800512
Anti-CD4 Mouse Monoclonal Antibody [clone: OTI5D9]
Antikörper
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR