Anti-CD4 Mouse Monoclonal Antibody [clone: OTI4H9]

Lieferant: OriGene
TA800522
ORIGTA800522EA 621 EUR
ORIGTA800522
Anti-CD4 Mouse Monoclonal Antibody [clone: OTI4H9]
Antikörper
  • Recommended Dilutions: Western Blot: 1:200-1:1000

Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR