Anti-PRR11 Mouse Monoclonal Antibody [clone: OTI1C1]

Lieferant: OriGene
TA800542
ORIGTA800542EA 621 EUR
ORIGTA800542
Anti-PRR11 Mouse Monoclonal Antibody [clone: OTI1C1]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: PRR11 (proline rich 11)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR