Anti-HBS1L Mouse Monoclonal Antibody [clone: OTI1C1]

Lieferant: OriGene
TA800554
ORIGTA800554EA 621 EUR
ORIGTA800554
Anti-HBS1L Mouse Monoclonal Antibody [clone: OTI1C1]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:500

Type: Primary
Antigen: HBS1L (HBS1-like (S. cerevisiae))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR