Anti-CCDC93 Mouse Monoclonal Antibody [clone: OTI2H4]

Lieferant: OriGene
TA800598
ORIGTA800598EA 621 EUR
ORIGTA800598
Anti-CCDC93 Mouse Monoclonal Antibody [clone: OTI2H4]
Antikörper
  • Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CCDC93 (coiled-coil domain containing 93)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR