Anti-SPN Mouse Monoclonal Antibody [clone: OTI6A4]

Lieferant: OriGene
TA800785
ORIGTA800785EA 621 EUR
ORIGTA800785
Anti-SPN Mouse Monoclonal Antibody [clone: OTI6A4]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: SPN (sialophorin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR