Anti-ABCB1 Mouse Monoclonal Antibody [clone: OTI2G4]

Lieferant: OriGene
TA800919
ORIGTA800919EA 621 EUR
ORIGTA800919
Anti-ABCB1 Mouse Monoclonal Antibody [clone: OTI2G4]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ABCB1 (ATP-binding cassette, sub-family B (MDR/TAP), member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR