Anti-TYMP Mouse Monoclonal Antibody [clone: OTI1G10]

Lieferant: OriGene
TA800961
ORIGTA800961EA 621 EUR
ORIGTA800961
Anti-TYMP Mouse Monoclonal Antibody [clone: OTI1G10]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TYMP (thymidine phosphorylase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR