Anti-CD99 Mouse Monoclonal Antibody [clone: OTI1F1]

Lieferant: OriGene
TA800963
ORIGTA800963EA 621 EUR
ORIGTA800963
Anti-CD99 Mouse Monoclonal Antibody [clone: OTI1F1]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CD99 (CD99 molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR