Anti-CD5 Mouse Monoclonal Antibody [clone: OTI9E9]

Lieferant: OriGene
TA802414
ORIGTA802414EA 621 EUR
ORIGTA802414
Anti-CD5 Mouse Monoclonal Antibody [clone: OTI9E9]
Antikörper
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: CD5 (CD5 molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR