Anti-TOP2A Mouse Monoclonal Antibody [clone: OTI1D5]

Lieferant: OriGene
TA802450
ORIGTA802450EA 621 EUR
ORIGTA802450
Anti-TOP2A Mouse Monoclonal Antibody [clone: OTI1D5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TOP2A (topoisomerase (DNA) II alpha 170kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR