Anti-RORB Mouse Monoclonal Antibody [clone: 4B7]

Lieferant: OriGene
TA806944
ORIGTA806944EA 621 EUR
ORIGTA806944
Anti-RORB Mouse Monoclonal Antibody [clone: 4B7]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: RORB
Clonality: Monoclonal
Clone: 4B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR