Anti-ABAT Mouse Monoclonal Antibody [clone: 1F10]

Lieferant: OriGene
TA806966
ORIGTA806966EA 621 EUR
ORIGTA806966
Anti-ABAT Mouse Monoclonal Antibody [clone: 1F10]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: ABAT
Clonality: Monoclonal
Clone: 1F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR