Anti-FLT4 Mouse Monoclonal Antibody [clone: 8C2]

Lieferant: OriGene
TA807002
ORIGTA807002EA 621 EUR
ORIGTA807002
Anti-FLT4 Mouse Monoclonal Antibody [clone: 8C2]
Antikörper
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: FLT4
Clonality: Monoclonal
Clone: 8C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR