Anti-EGFR Mouse Monoclonal Antibody [clone: UMAB95]

Lieferant: OriGene
UM500070
ORIGUM500070EA 847 EUR
ORIGUM500070
Anti-EGFR Mouse Monoclonal Antibody [clone: UMAB95]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:100

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: UMAB95
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR