Anti-VIM Mouse Monoclonal Antibody [clone: UMAB159]

Lieferant: OriGene
UM800054
ORIGUM800054EA 847 EUR
ORIGUM800054
Anti-VIM Mouse Monoclonal Antibody [clone: UMAB159]
Antikörper
Recommended Dilutions: Immunochemistry: 1:200

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: UMAB159
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR