Culture media in bags, ReadyBags™
Lieferant: THERMO FISHER DIAGNOSTICS
BM0104T
BM0204T
BM1104T
BM9901A
OXOIBM0104TEA
225
EUR
OXOIBM0104T
OXOIBM0204T
OXOIBM1104T
OXOIBM9901A
Culture media in bags, ReadyBags™
Medien für die Mikrobiologie
Fertigmedien für die Mikrobiologie
ReadyBags™ are designed for use in food testing by laboratories with large numbers of samples requiring large volumes of broth media or diluent. Each ReadyBag™ contains 3 liters and is intended for use with an automatic dispenser. The ReadyBag™ is connected to tubing for standard laboratory equipment, such as gravimetric diluters or peristaltic pumps, by means of a connector. Connectors are available in two formats: autoclavable, re-usable stainless steel connectors or sterile, plastic, single-use connectors.
- For large numbers of samples
- Available in 3 L
- Once empty, the bag can be disposed as normal laboratory waste