Culture media in bags, Dry-Bags
Lieferant: THERMO FISHER DIAGNOSTICS
DB0509M DB0733M DB1049M DB0895V DB1049W
OXOIDB0509MEA 1460 EUR
OXOIDB0509M OXOIDB0733M OXOIDB1049M OXOIDB0895V OXOIDB1049W
Culture media in bags, Dry-Bags
Medien für die Mikrobiologie Fertigmedien für die Mikrobiologie
Dry-Bags take the hassle out of media preparation. They remove the need for glassware, making them a safe and convenient option for food-testing laboratories and eliminate the need for washing of glassware and heat sterilization. Furthermore, there is no need to wait for hot media to cool down. Once water is added and powder dissolved, Dry-Bags are ready to use immediately. When empty, the bag is simply disposed of with normal laboratory waste.
  • Dry-Bag is irradiated for sterility
  • Available with filters attached for convenience or without filters as a lower-cost option
  • Dry-Bags supplied without filters, autoclavable, reusable filters are also available