Anti-STUB1 Rabbit Polyclonal Antibody

Lieferant: Merck Millipore (Oncogene)
PC711-100
PC711-100EA 586 EUR
PC711-100
Anti-STUB1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STUB1 (STIP1 homology and U-box containing protein 1, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR