Substrate for western blotting, 1-Step™ NBT/BCIP
Lieferant: Thermo Fisher Scientific
34042
PIER34042EA
266
EUR
PIER34042
Substrate for western blotting, 1-Step™ NBT/BCIP
Elektrophorese-Reagenzien
Western Blotting Reagents
The 1-Step NBT/BCIP is a one-component sensitive precipitating alkaline phosphatase substrate for immunoblotting, immunohistochemistry, and in situ hybridisation applications.