DAB substrate kit, metal enhanced, Pierce™
Lieferant: Thermo Fisher Scientific
34065
PIER34065EA
298
EUR
PIER34065
DAB substrate kit, metal enhanced, Pierce™
Elektrophorese-Reagenzien
Western Blotting Reagents
The Pierce™ Metal enhanced DAB substrate kit uses cobalt and nickel chloride in a special formulation of diaminobenzidine peroxidase substrate that yields intense colour for immunohistochemical staining.
- Up to 50 times more sensitive than traditional DAB methods
- Low background and high intensity
- Only 2 components
- Working solution stable for more than six hours at room temperature