Fluorescent western blot stripping buffer, Restore™
Lieferant: Thermo Fisher Scientific
62299 62300
PIER62299EA 59 EUR
PIER62299 PIER62300
Fluorescent western blot stripping buffer, Restore™
Elektrophorese-Reagenzien Western Blotting Reagents
Restore™ Fluorescent Western Blot Stripping Buffer is a gentle and highly effective reagent for quickly removing primary and near-infrared (IR) dye-labeled secondary antibodies from western blots.
  • Strip blots in only 15 minutes at room temperature
  • Reprobe the same PVDF membrane for multiple targets
  • Effectively strips blots the first time
  • No need to run new gels and prepare a new blot