Dialyseeinheiten, Float-A-Lyzer™
Lieferant: Thermo Fisher Scientific
87761 87762 87760 87763
PIER87761EA 412 EUR
PIER87761 PIER87762 PIER87760 PIER87763
Dialyseeinheiten, Float-A-Lyzer™
Schläuche Dialyseschläuche
These dialysis flasks facilitate simple and effective removal of buffer salts and small contaminants from proteins and other macromolecules in sample volumes up to 250 ml.
  • Easy to use
  • Color-coded for easy identification
  • No leakage, sample remains intact with no contamination
  • High surface area to volume ratio maximizes diffusion rate
Devices are constructed from two sheets of low protein-binding, regenerated cellulose membranes to ensure maximum sample recovery and purity and contain up to 85% less plastic per volume compared to other devices. Slide-A-Lyzer dialysis flasks are compatible with most laboratory buffers and can accomplish low-molecular weight contaminant removal, buffer exchange and desalting in 8 hours to overnight.