Pierce™ Biotin 3' End DNA Labelling Kit
Lieferant: Thermo Fisher Scientific
89818
PIER89818EA
628
EUR
PIER89818
Pierce™ Biotin 3' End DNA Labelling Kit
Elektrophorese-Reagenzien
Western Blotting Reagents
This Biotin 3' End DNA Labelling Kit is used for tagging single-stranded DNA primers with biotin for use in non radioactive electrophoretic mobility shift assays (EMSA) and other nucleic acid detection methods.
- Non isotopic labelling eliminates the hassle of hazardous radioactive materials or difficult-to-dispose-of waste
- 1-3 biotinylated ribonucleotides onto the 3' end of DNA strands for less interference with hybridisation or sequence-specific binding of proteins
- Biotin-labeled probes are stable for more than one year
- Fast, efficient 30 minute labelling procedure
Lieferumfang: The kit contains sufficient reagents for 20 reactions (each with 5 pmol DNA) and includes 5X TdT Reaction Buffer,1 ml; TdT (terminal deoxynucleotidyl tranferase), 5 µl; Biotin-11-UTP,1 µl; unlabeled control oligo,14 µl; and biotin-control oligo,4 µl.