Pierce™ In-solution Tryptic Digestion and Guanidination Kit
Lieferant: Thermo Fisher Scientific
The In-Solution Tryptic Digestion and Guanidination Kit contains proteomics grade trypsin, buffers and reagents to alkylate and digest proteins and then guanidinate the peptide fragments for mass spectrometry.
- Optimised trypsin digestion producing complete digests of protein ranging from 0,025 to 10 µg
- Optimised guanidination where reaction produces little or no N-terminal modification
- Protein can be digested and guanidinated all in one day
Accurate identification of proteins and analysis of post-translational modifications by mass spectrometry require accurate and complete protein digestion and peptide modification and this kit provides an optimised procedure and reagents for approximately 90 digests.