S-Nitrosylation western blot kit, Pierce™
Lieferant: Thermo Fisher Scientific
90105 90106
PIER90105EA 546 EUR
PIER90105 PIER90106
S-Nitrosylation western blot kit, Pierce™
Elektrophorese-Reagenzien Western Blotting Reagents
The Pierce™ S-Nitrosylation Western Blot Kit enables sensitive detection of protein S-nitrosocysteine post-translational modifications.
  • Optimised labeling reactions switch S-nitrosocysteine post translational modifications with iodoTMT™ reagent for easy detection by Western blot using anti-TMT™ antibody
  • Uses a non-biological iodoTMT™ reagent for labeling instead of HPDP-biotin, resulting in less background during Western blot detection
  • Labeling method is compatible with alternative methods of analysis, including enrichment via anti-TMT™ resin and multiplex quantitation by mass spectrometry
  • All needed reagents included one kit
The Pierce™ S-Nitrosylation Western Blot Kit contains a cell lysis/reaction buffer, a sulfhydryl-reactive blocking agent, a reducing agent, a labeling agent and a detection monoclonal antibody. Unmodified cysteines are first blocked using a sulfhydryl-reactive compound (MMTS). S-nitrosylated cysteines are then selectively reduced with ascorbate in HENS Buffer for specific labeling with iodoTMTzero™ reagents, which irreversibly bind to the cysteine thiol that was S-nitrosylated. Detection of the TMT™ reagent-modified proteins is facilitated using an anti-TMT™ antibody.
Lieferumfang: The kit includes HENS Buffer (100 ml), methyl methanethiosulfonat (MMTS, 200 mg), sodium ascorbate (20 mg), iodoTMTzero™ label reagent (2×0,2 mg) and anti-TMT™ antibody (100 µl).