Anti-BYM338 Recombinant Antibody

Lieferant: ProSci Inc.
10-586
PRSI10-586EA 654 EUR
PRSI10-586
Anti-BYM338 Recombinant Antibody
Antikörper
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2-lambda
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR