Human PAK4 peptide (13 aa in the center)
Lieferant: ProSci Inc.
3077P
PRSI3077PEA 122 EUR
PRSI3077P
Human PAK4 peptide (13 aa in the center)
Proteine und Peptide
PAK4 peptide is used for blocking activity of PAK4 antibody.